1'-Boc-spiro[indene-1,4'-piperidine]

Lieferant: Apollo Scientific

Synonyme: tert-Butyl spiro[indene-1,4'-piperidine]-1'-carboxylate

OR470774-5G
APOSOR470774-5GEA 567 EUR
APOSOR470774-5G
1'-Boc-spiro[indene-1,4'-piperidine]
1'-Boc-spiro[indene-1,4'-piperidine]
Formel: C₁₈H₂₃NO₂
Molecular Weight: 285,381 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03160565
CAS-Nummer: 137419-24-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR