5-Bromo-2-methylquinoline

Lieferant: Apollo Scientific

OR470776-1G OR470776-250MG OR470776-5G
APOSOR470776-1GEA 184 EUR
APOSOR470776-1G APOSOR470776-250MG APOSOR470776-5G
5-Bromo-2-methylquinoline
5-Bromo-2-methylquinoline
Formel: C₁₀H₈BrN
Molecular Weight: 222.081 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13189490
CAS-Nummer: 54408-52-5

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR