2-Chloro-3-phenylpyrazine

Lieferant: Apollo Scientific

OR470777-1G
APOSOR470777-1GEA 1120 EUR
APOSOR470777-1G
2-Chloro-3-phenylpyrazine
2-Chloro-3-phenylpyrazine
Formel: C₁₀H₇ClN₂
Molecular Weight: 190,629 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01411765
CAS-Nummer: 41270-65-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR