3-(Iodomethyl)pyridine hydroiodide
Lieferant: Apollo Scientific
OR470780-5G
APOSOR470780-5GEA
601
EUR
APOSOR470780-5G
3-(Iodomethyl)pyridine hydroiodide
3-(Iodomethyl)pyridine hydroiodide
Formel:
C₆H₆IN·HI Molecular Weight: 346,935 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01321315 CAS-Nummer: 69966-59-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...