2-(2-Iodoethyl)-2-methyl-1,3-dioxolane

Lieferant: Apollo Scientific

OR470781-1G
APOSOR470781-1GEA 804 EUR
APOSOR470781-1G
2-(2-Iodoethyl)-2-methyl-1,3-dioxolane
2-(2-Iodoethyl)-2-methyl-1,3-dioxolane
Formel: C₆H₁₁IO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD27500653
CAS-Nummer: 53750-51-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR