6-[Bis(Boc)amino]purine

Lieferant: Apollo Scientific

Synonyme: tert-Butyl N-tert-butoxycarbonyl-N-(7H-purin-6-yl)carbamate

OR470783-5G
APOSOR470783-5GEA 114 EUR
APOSOR470783-5G
6-[Bis(Boc)amino]purine
6-[Bis(Boc)amino]purine
Formel: C₁₅H₂₁N₅O₄
Molecular Weight: 335,358 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD17676144
CAS-Nummer: 309947-86-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR