(S)-6,6'-Dibromo-2,2'-dimethoxy-1,1'-binaphthalene

Lieferant: Apollo Scientific

OR470785-5G
APOSOR470785-5GEA 528 EUR
APOSOR470785-5G
(S)-6,6'-Dibromo-2,2'-dimethoxy-1,1'-binaphthalene
(S)-6,6'-Dibromo-2,2'-dimethoxy-1,1'-binaphthalene
Formel: C₂₂H₁₆Br₂O₂
Molecular Weight: 472.169 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06654379
CAS-Nummer: 117745-41-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR