2-[3-(Cyclopropylmethoxy)-4-ethoxyphenyl]acetic acid
Lieferant: Apollo Scientific
OR470787-5G
APOSOR470787-5GEA
916
EUR
APOSOR470787-5G
2-[3-(Cyclopropylmethoxy)-4-ethoxyphenyl]acetic acid
2-[3-(Cyclopropylmethoxy)-4-ethoxyphenyl]acetic acid
Formel:
C₁₄H₁₈O₄ Molecular Weight: 250,29 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD27500650 CAS-Nummer: 2006277-02-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...