2-(4-Ethoxy-3-isopropoxyphenyl)acetic acid

Lieferant: Apollo Scientific

OR470788-5G
APOSOR470788-5GEA 916 EUR
APOSOR470788-5G
2-(4-Ethoxy-3-isopropoxyphenyl)acetic acid
2-(4-Ethoxy-3-isopropoxyphenyl)acetic acid
Formel: C₁₃H₁₈O₄
Molecular Weight: 238,28 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD27500649
CAS-Nummer: 2006277-26-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR