2-[3-(Benzyloxy)-4-ethoxyphenyl]acetic acid

Lieferant: Apollo Scientific

OR470789-5G
APOSOR470789-5GEA 916 EUR
APOSOR470789-5G
2-[3-(Benzyloxy)-4-ethoxyphenyl]acetic acid
2-[3-(Benzyloxy)-4-ethoxyphenyl]acetic acid
Formel: C₁₇H₁₈O₄
Molecular Weight: 286,322 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06208482
CAS-Nummer: 85475-84-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR