2-(3-Isopropoxy-4-methoxyphenyl)acetic acid
Lieferant: Apollo Scientific
OR470791-5G
APOSOR470791-5GEA
916
EUR
APOSOR470791-5G
2-(3-Isopropoxy-4-methoxyphenyl)acetic acid
2-(3-Isopropoxy-4-methoxyphenyl)acetic acid
Formel:
C₁₂H₁₆O₄ Lagertemperatur: Raumtemperatur |
MDL:
MFCD27500647 CAS-Nummer: 1823817-32-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...