2-[3-(Benzyloxy)-4-methoxyphenyl]acetic acid

Lieferant: Apollo Scientific

OR470792-5G
APOSOR470792-5GEA 713 EUR
APOSOR470792-5G
2-[3-(Benzyloxy)-4-methoxyphenyl]acetic acid
2-[3-(Benzyloxy)-4-methoxyphenyl]acetic acid
Formel: C₁₆H₁₆O₄
Lagertemperatur: Raumtemperatur
MDL: MFCD00183334
CAS-Nummer: 5487-33-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR