2-[3-(tert-Butoxy)-4-methoxyphenyl]acetic acid

Lieferant: Apollo Scientific

OR470793-1G OR470793-5G
APOSOR470793-1GEA 397 EUR
APOSOR470793-1G APOSOR470793-5G
2-[3-(tert-Butoxy)-4-methoxyphenyl]acetic acid
2-[3-(tert-Butoxy)-4-methoxyphenyl]acetic acid
Formel: C₁₃H₁₈O₄
Lagertemperatur: Raumtemperatur
MDL: MFCD27500646
CAS-Nummer: 2006277-90-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR