4-(Cyclopropylmethoxy)-3-methylbenzoic acid
Lieferant: Apollo Scientific
OR470794-5G
APOSOR470794-5GEA
804
EUR
APOSOR470794-5G
4-(Cyclopropylmethoxy)-3-methylbenzoic acid
4-(Cyclopropylmethoxy)-3-methylbenzoic acid
Formel:
C₁₂H₁₄O₃ Molecular Weight: 206,238 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD16852894 CAS-Nummer: 1252903-16-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...