4-Isopropoxy-3-methylbenzoic acid

Lieferant: Apollo Scientific

OR470795-5G
APOSOR470795-5GEA 804 EUR
APOSOR470795-5G
4-Isopropoxy-3-methylbenzoic acid
4-Isopropoxy-3-methylbenzoic acid
Formel: C₁₁H₁₄O₃
Molecular Weight: 194,227 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD14535425
CAS-Nummer: 856165-81-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR