4-Ethoxy-3-methylbenzoic acid
Lieferant: Apollo Scientific
OR470796-5G
APOSOR470796-5GEA
601
EUR
APOSOR470796-5G
4-Ethoxy-3-methylbenzoic acid
4-Ethoxy-3-methylbenzoic acid
Formel:
C₁₀H₁₂O₃ Molecular Weight: 180.2 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD13920122 CAS-Nummer: 92315-60-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...