N-Isobutylcyclopropanamine

Lieferant: Apollo Scientific

OR470797-1G OR470797-5G
APOSOR470797-1GEA 397 EUR
APOSOR470797-1G APOSOR470797-5G
N-Isobutylcyclopropanamine
N-Isobutylcyclopropanamine
Formel: C₇H₁₅N
Lagertemperatur: Raumtemperatur
MDL: MFCD09047821
CAS-Nummer: 215523-02-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR