2-(Phenylamino)propanenitrile

Lieferant: Apollo Scientific

OR470798-5G
APOSOR470798-5GEA 619 EUR
APOSOR470798-5G
2-(Phenylamino)propanenitrile
2-(Phenylamino)propanenitrile
Formel: C₉H₁₀N₂
Molecular Weight: 146,189 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12816738
CAS-Nummer: 2182-39-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR