2-Aminoimidazole-4-carboxylic acid

Lieferant: Apollo Scientific

OR470799-1G
APOSOR470799-1GEA 1330 EUR
APOSOR470799-1G
2-Aminoimidazole-4-carboxylic acid
2-Aminoimidazole-4-carboxylic acid
Formel: C₄H₅N₃O₂
Molecular Weight: 127,101 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08668249
CAS-Nummer: 860011-60-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR