3-Bromo-7-chloro-1H-pyrazolo[4,3-d]pyrimidine
Lieferant: Apollo Scientific
OR470800-1G
APOSOR470800-1GEA
1090
EUR
APOSOR470800-1G
3-Bromo-7-chloro-1H-pyrazolo[4,3-d]pyrimidine
3-Bromo-7-chloro-1H-pyrazolo[4,3-d]pyrimidine
Formel:
C₅H₂BrClN₄ Molecular Weight: 233,453 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09746269 CAS-Nummer: 68510-70-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...