Benzyl (S)-3-(Boc-amino)-4-iodobutanoate

Lieferant: Apollo Scientific

OR470801-1G
APOSOR470801-1GEA 1090 EUR
APOSOR470801-1G
Benzyl (S)-3-(Boc-amino)-4-iodobutanoate
Benzyl (S)-3-(Boc-amino)-4-iodobutanoate
Formel: C₁₆H₂₂INO₄
Molecular Weight: 419,255 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD27500657
CAS-Nummer: 161529-22-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR