tert-Butyl (S)-3-(Boc-amino)-4-bromobutanoate

Lieferant: Apollo Scientific

OR470802-1G
APOSOR470802-1GEA 804 EUR
APOSOR470802-1G
tert-Butyl (S)-3-(Boc-amino)-4-bromobutanoate
tert-Butyl (S)-3-(Boc-amino)-4-bromobutanoate
Formel: C₁₃H₂₄BrNO₄
Molecular Weight: 338,238 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD27500656
CAS-Nummer: 2006286-94-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR