tert-Butyl (S)-3-(Boc-amino)-4-iodobutanoate

Lieferant: Apollo Scientific

OR470803-1G
APOSOR470803-1GEA 804 EUR
APOSOR470803-1G
tert-Butyl (S)-3-(Boc-amino)-4-iodobutanoate
tert-Butyl (S)-3-(Boc-amino)-4-iodobutanoate
Formel: C₁₃H₂₄INO₄
Molecular Weight: 385,238 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD27500655
CAS-Nummer: 1802736-56-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR