5-(4-Hydroxyphenyl)thiophene-2-carboxylic acid
Lieferant: Apollo Scientific
OR470805-1G
APOSOR470805-1GEA
601
EUR
APOSOR470805-1G
5-(4-Hydroxyphenyl)thiophene-2-carboxylic acid
5-(4-Hydroxyphenyl)thiophene-2-carboxylic acid
Formel:
C₁₁H₈O₃S Molecular Weight: 220,244 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD27500654 CAS-Nummer: 116016-57-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...