5-(4-Bromophenyl)-3-methylisoxazole

Lieferant: Apollo Scientific

OR470806-1G
APOSOR470806-1GEA 499 EUR
APOSOR470806-1G
5-(4-Bromophenyl)-3-methylisoxazole
5-(4-Bromophenyl)-3-methylisoxazole
Formel: C₁₀H₈BrNO
Molecular Weight: 238,081 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09756424
CAS-Nummer: 52063-43-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR