4-Chloro-3-methylbutanoyl chloride
Lieferant: Apollo Scientific
OR470807-1G
APOSOR470807-1GEA
713
EUR
APOSOR470807-1G
4-Chloro-3-methylbutanoyl chloride
4-Chloro-3-methylbutanoyl chloride
Formel:
C₅H₈Cl₂O Molecular Weight: 155,022 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD19232353 CAS-Nummer: 63480-13-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...