4-Chloro-2-methylbutanoyl chloride

Lieferant: Apollo Scientific

OR470808-5G
APOSOR470808-5GEA 0 EUR
APOSOR470808-5G
4-Chloro-2-methylbutanoyl chloride
4-Chloro-2-methylbutanoyl chloride
Formel: C₅H₈Cl₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD19232042
CAS-Nummer: 51708-72-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR