2-(Bromomethyl)-7-methoxynaphthalene

Lieferant: Apollo Scientific

OR470813-1G
APOSOR470813-1GEA 1420 EUR
APOSOR470813-1G
2-(Bromomethyl)-7-methoxynaphthalene
2-(Bromomethyl)-7-methoxynaphthalene
Formel: C₁₂H₁₁BrO
Molecular Weight: 251,119 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD18410675
CAS-Nummer: 103143-23-3
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR