2-Methoxy-7-methylnaphthalene

Lieferant: Apollo Scientific

OR470814-1G
APOSOR470814-1GEA 249 EUR
APOSOR470814-1G
2-Methoxy-7-methylnaphthalene
2-Methoxy-7-methylnaphthalene
Formel: C₁₂H₁₂O
Molecular Weight: 172,223 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18412839
CAS-Nummer: 1131-53-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR