N-Cyclohexyl-1-imidazolecarboxamide

Lieferant: Apollo Scientific

OR470816-5G
APOSOR470816-5GEA 804 EUR
APOSOR470816-5G
N-Cyclohexyl-1-imidazolecarboxamide
N-Cyclohexyl-1-imidazolecarboxamide
Formel: C₁₀H₁₅N₃O
Lagertemperatur: Raumtemperatur
MDL: MFCD01175643
CAS-Nummer: 91977-33-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR