N-Cyclopentyl-1-imidazolecarboxamide

Lieferant: Apollo Scientific

OR470817-5G
APOSOR470817-5GEA 804 EUR
APOSOR470817-5G
N-Cyclopentyl-1-imidazolecarboxamide
N-Cyclopentyl-1-imidazolecarboxamide
Formel: C₉H₁₃N₃O
Molecular Weight: 179,219 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11099393
CAS-Nummer: 154696-60-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR