N-Isobutyl-1-imidazolecarboxamide

Lieferant: Apollo Scientific

OR470818-5G
APOSOR470818-5GEA 713 EUR
APOSOR470818-5G
N-Isobutyl-1-imidazolecarboxamide
N-Isobutyl-1-imidazolecarboxamide
Formel: C₈H₁₃N₃O
Molecular Weight: 167,208 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11099706
CAS-Nummer: 149047-76-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR