N-Ethyl-1-imidazolecarboxamide

Lieferant: Apollo Scientific

OR470819-5G
APOSOR470819-5GEA 713 EUR
APOSOR470819-5G
N-Ethyl-1-imidazolecarboxamide
N-Ethyl-1-imidazolecarboxamide
Formel: C₆H₉N₃O
Molecular Weight: 139,155 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11099735
CAS-Nummer: 58124-84-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR