3,3-Di(hydroxymethyl)azetidine
Lieferant: Apollo Scientific
OR470820-1G
APOSOR470820-1GEA
1010
EUR
APOSOR470820-1G
3,3-Di(hydroxymethyl)azetidine
3,3-Di(hydroxymethyl)azetidine
Formel:
C₅H₁₁NO₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD19217762 CAS-Nummer: 45512-27-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...