Ethyl 6-Bromo-5-methoxyindole-2-carboxylate

Lieferant: Apollo Scientific

OR470821-1G
APOSOR470821-1GEA 1010 EUR
APOSOR470821-1G
Ethyl 6-Bromo-5-methoxyindole-2-carboxylate
Ethyl 6-Bromo-5-methoxyindole-2-carboxylate
Formel: C₁₂H₁₂BrNO₃
Molecular Weight: 298,133 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD20483268
CAS-Nummer: 128593-40-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR