Methyl 5-[4-(ethoxycarbonyl)phenyl]isoxazole-3-carboxylate

Lieferant: Apollo Scientific

OR470822-1G
APOSOR470822-1GEA 397 EUR
APOSOR470822-1G
Methyl 5-[4-(ethoxycarbonyl)phenyl]isoxazole-3-carboxylate
Methyl 5-[4-(ethoxycarbonyl)phenyl]isoxazole-3-carboxylate
Formel: C₁₄H₁₃NO₅
Molecular Weight: 275,257 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD27935378
CAS-Nummer: 2006277-68-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR