2-Amino-5-methoxy-1-methylbenzimidazole

Lieferant: Apollo Scientific

OR470826-1G
APOSOR470826-1GEA 794 EUR
APOSOR470826-1G
2-Amino-5-methoxy-1-methylbenzimidazole
2-Amino-5-methoxy-1-methylbenzimidazole
Formel: C₉H₁₁N₃O
Molecular Weight: 177,2 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01658296
CAS-Nummer: 1805-02-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR