2-Amino-6-methoxy-1-methylbenzimidazole
Lieferant: Apollo Scientific
OR470827-1G
APOSOR470827-1GEA
0
EUR
APOSOR470827-1G
2-Amino-6-methoxy-1-methylbenzimidazole
2-Amino-6-methoxy-1-methylbenzimidazole
Formel:
C₉H₁₁N₃O Molecular Weight: 177,203 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD23904053 CAS-Nummer: 945023-35-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...