2-Amino-1,7-dimethylbenzimidazole

Lieferant: Apollo Scientific

Synonyme: 1,7-Dimethyl-1H-benzo[d]imidazol-2-amine

OR470828-1G
APOSOR470828-1GEA 794 EUR
APOSOR470828-1G
2-Amino-1,7-dimethylbenzimidazole
2-Amino-1,7-dimethylbenzimidazole
Formel: C₉H₁₁N₃
Molecular Weight: 161,204 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03725798
CAS-Nummer: 945021-49-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR