2-Amino-4-methoxy-1-methylbenzimidazole
Lieferant: Apollo Scientific
OR470829-1G
APOSOR470829-1GEA
794
EUR
APOSOR470829-1G
2-Amino-4-methoxy-1-methylbenzimidazole
2-Amino-4-methoxy-1-methylbenzimidazole
Formel:
C₉H₁₁N₃O Molecular Weight: 177,203 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD18825732 CAS-Nummer: 33235-33-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...