N-Methylpiperidine-3-carboxamide

Lieferant: Apollo Scientific

OR470831-5G
APOSOR470831-5GEA 947 EUR
APOSOR470831-5G
N-Methylpiperidine-3-carboxamide
N-Methylpiperidine-3-carboxamide
Formel: C₇H₁₄N₂O
Molecular Weight: 142,199 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06408743
CAS-Nummer: 5115-98-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR