1-Oxa-6-azaspiro[3.5]nonane

Lieferant: Apollo Scientific

OR470832-1G
APOSOR470832-1GEA 1420 EUR
APOSOR470832-1G
1-Oxa-6-azaspiro[3.5]nonane
1-Oxa-6-azaspiro[3.5]nonane
Formel: C₇H₁₃NO
Lagertemperatur: Raumtemperatur
MDL: MFCD13180593
CAS-Nummer: 204388-70-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR