(S)-2-Amino-4-(7-hydroxy-2-oxo-2H-chromen-4-yl)butanoic acid

Lieferant: Apollo Scientific

Synonyme: H-4-(7-Hydroxy-4-coumarinyl)-Abu-OH, 7-HCAA

OR470836-1G
APOSOR470836-1GEA 2450 EUR
APOSOR470836-1G
(S)-2-Amino-4-(7-hydroxy-2-oxo-2H-chromen-4-yl)butanoic acid
(S)-2-Amino-4-(7-hydroxy-2-oxo-2H-chromen-4-yl)butanoic acid
Formel: C₁₃H₁₃NO₅
Molecular Weight: 263,249 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22377747
CAS-Nummer: 905442-42-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR