3-Bromo-2-chlorophenyl Diethylcarbamate

Lieferant: Apollo Scientific

OR470837-5G
APOSOR470837-5GEA 448 EUR
APOSOR470837-5G
3-Bromo-2-chlorophenyl Diethylcarbamate
3-Bromo-2-chlorophenyl Diethylcarbamate
Formel: C₁₁H₁₃BrClNO₂
Molecular Weight: 306,583 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08166318
CAS-Nummer: 863870-81-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR