3-Bromophenyl Diethylcarbamate

Lieferant: Apollo Scientific

Synonyme: Carbamic acid, N,N-diethyl-, 3-bromophenyl ester

OR470838-5G
APOSOR470838-5GEA 367 EUR
APOSOR470838-5G
3-Bromophenyl Diethylcarbamate
3-Bromophenyl Diethylcarbamate
Formel: C₁₁H₁₄BrNO₂
Molecular Weight: 272,138 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD27930373
CAS-Nummer: 863870-72-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR