Benzyl 1-Acetylazetidine-3-carboxylate
Lieferant: Apollo Scientific
OR470839-5G
APOSOR470839-5GEA
1120
EUR
APOSOR470839-5G
Benzyl 1-Acetylazetidine-3-carboxylate
Benzyl 1-Acetylazetidine-3-carboxylate
Formel:
C₁₃H₁₅NO₃ Molecular Weight: 233,263 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD27935374 CAS-Nummer: 1823224-94-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...