2-Bromo-4-methyl-5-nitroanisole

Lieferant: Apollo Scientific

OR470842-5G
APOSOR470842-5GEA 289 EUR
APOSOR470842-5G
2-Bromo-4-methyl-5-nitroanisole
2-Bromo-4-methyl-5-nitroanisole
Formel: C₈H₈BrNO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD27935373
CAS-Nummer: 41447-19-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR