Ethyl 2-Methyltetrahydrofuran-2-carboxylate

Lieferant: Apollo Scientific

OR470844-5G
APOSOR470844-5GEA 916 EUR
APOSOR470844-5G
Ethyl 2-Methyltetrahydrofuran-2-carboxylate
Ethyl 2-Methyltetrahydrofuran-2-carboxylate
Formel: C₈H₁₄O₃
Molecular Weight: 158,195 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD23847310
CAS-Nummer: 1540337-79-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR