8-Benzyl-1,8-diazaspiro[5.5]undecane

Lieferant: Apollo Scientific

OR470846-1G
APOSOR470846-1GEA 1420 EUR
APOSOR470846-1G
8-Benzyl-1,8-diazaspiro[5.5]undecane
8-Benzyl-1,8-diazaspiro[5.5]undecane
Formel: C₁₆H₂₄N₂
Lagertemperatur: Raumtemperatur
MDL: MFCD11223532
CAS-Nummer: 1086395-04-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR