2-Cbz-2,8-diazaspiro[5.5]undecane

Lieferant: Apollo Scientific

OR470847-1G
APOSOR470847-1GEA 1540 EUR
APOSOR470847-1G
2-Cbz-2,8-diazaspiro[5.5]undecane
2-Cbz-2,8-diazaspiro[5.5]undecane
Formel: C₁₇H₂₄N₂O₂
Molecular Weight: 288,385 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11223514
CAS-Nummer: 1086394-63-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR